TJ-1L-BK
Anti-S (clone MS-94), Blood Grouping Reagent
BIOSCOT™ Anti-S from the clone MS-94, Human IgM, Blood Grouping Reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
human
Quality Level
antibody form
unpurified
clone
MS-94, monoclonal
form
liquid
packaging
pkg of 1 L
manufacturer/tradename
BIOSCOT™
concentration
60-80 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
7.1-7.3
isotype
IgM
shipped in
ambient
storage temp.
2-8°C
General description
Contains 0.1% (w/v) Sodium Azide
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months from date of filtration
Product formulation conforms to the requirements of the EU IVD directive.
For In Vitro use only
Filtered through a 0.22μm terminal filter
Shelf Life: 36 months from date of filtration
Product formulation conforms to the requirements of the EU IVD directive.
For In Vitro use only
Application
Bulk
For use by Tube;Slide;Microplate techniques
Legal Information
BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service