Skip to Content
Merck

Skip To

SYMBRACK2

Steritest® racks

to hold-up to 4 Steritest®carrying trays, for use with Steritest® NEO Devices, pkg of 1 kit

Synonym(s):

Tray holder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
23151818
NACRES:
NB.24
eCl@ss:
32014001
Manufacturer/tradename:
Steritest®
Material:
stainless steel
Packaging:
pkg of 1 kit
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

stainless steel

agency

EP 2.6.1, JP 4.06, USP 71

packaging

pkg of 1 kit

manufacturer/tradename

Steritest®

input

sample type pharmaceutical(s)

application(s)

pharmaceutical
sample preparation
sterility testing

compatibility

for use with Steritest® NEO Devices, for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01), for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01), for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01), for use with Steritest® Symbio Pump, standard for VHP (Autoclavable)

shipped in

ambient

General description

The Steritest® rack can only be used together with the Steritest® carrying trays. The combination of 4 Steritest® carrying trays and a Steritest® rack enables the user to simultaneously and safely transport 20 media-filled canisters. The Steritest® carrying trays and Steritest® rack can remain in the incubator.

Application

Steritest® racks are used to streamline the workflow and increase safety during laboratory filtration and sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SYMBCAN08SYMBSVB01SYMBDSF02
manufacturer/tradename

Steritest®

manufacturer/tradename

Steritest®

manufacturer/tradename

Steritest®

manufacturer/tradename

Steritest®

material

stainless steel

material

polyphenylsulfone (PPSU)

material

316L stainless steel

material

316L stainless steel support (drain tray), EPDM rubber seal

packaging

pkg of 1 kit

packaging

pkg of (8 trays per pack)

packaging

pkg of 1 unit

packaging

-

agency

EP 2.6.1, USP 71, JP 4.06

agency

EP 2.6.1, JP 4.06, USP 71

agency

EP (2.6.1), JP (4.06), USP (71)

agency

EP (2.6.1), JP (4.06), USP (71)

input

sample type pharmaceutical(s)

input

sample type pharmaceutical(s)

input

pharmaceutical(s)

input

pharmaceutical(s)

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters