Skip to Content
Merck
All Photos(2)

Documents

SYMBCBH01

Millipore

Steritest® Symbio bottle holder basket

With fastening screw, for use with Steritest® Symbio pump

Synonym(s):

Bottle mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

material

316L stainless steel
polyphenylsulfone (PPSU) (Fastening Screw)

Agency

EP (2.6.1)
JP (4.06)
USP (71)

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® Symbio bottle holder basket with its fastening screw and clip.

Application

Accessories for Streamlining Your Workflow and Increasing Safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service