Skip to Content
Merck
All Photos(1)

Key Documents

SX1247

Supelco

Sulfuric acid

95.5-96.5%, OmniTrace

Synonym(s):

Sulfuric acid, Oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-639-5

Product Name

Sulfuric Acid 95.5-96.5%, OmniTrace, for inorganic trace analysis

grade

for inorganic trace analysis

Quality Level

Agency

suitable for EPA 300

vapor density

<0.3 (25 °C, vs air)

vapor pressure

0.0001 hPa ( 20 °C)
1 mmHg ( 146 °C)

Assay

93-98%

form

liquid

pH

0.3 (25 °C, 49 g/L in H2O)

bp

~290 °C (lit.)

mp

-20 °C

density

1.840 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.7 ppm
nitrate (NO3-): ≤0.2 ppm

cation traces

Ag: ≤1 ppb
Al: ≤1 ppb
As: ≤0.5 ppb
Au: ≤0.5 ppb
Ba: ≤0.1 ppb
Be: ≤0.1 ppb
Bi: ≤0.1 ppb
Ca: ≤1 ppb
Cd: ≤0.5 ppb
Ce: ≤0.1 ppb
Co: ≤0.5 ppb
Cr: ≤0.5 ppb
Cs: ≤0.1 ppb
Cu: ≤0.5 ppb
Dy: ≤0.1 ppb
Er: ≤0.1 ppb
Eu: ≤0.1 ppb
Fe: ≤1 ppb
Ga: ≤0.1 ppb
Gd: ≤0.1 ppb
Ge: ≤1 ppb
Hf: ≤0.1 ppb
Hg: ≤0.1 ppb
Ho: ≤0.1 ppb
In: ≤0.1 ppb
K: ≤1 ppb
La: ≤0.1 ppb
Li: ≤0.5 ppb
Lu: ≤0.1 ppb
Mg: ≤1 ppb
Mn: ≤0.5 ppb
Mo: ≤0.5 ppb
Na: ≤1 ppb
Nb: ≤0.1 ppb
Nd: ≤0.1 ppb
Ni: ≤0.5 ppb
P: ≤0.05 ppm
Pb: ≤0.1 ppb
Pr: ≤0.1 ppb
Rb: ≤0.5 ppb
Rh: ≤0.5 ppb
Sb: ≤1 ppb
Sc: ≤0.1 ppb
Se: ≤10 ppb
Sm: ≤0.1 ppb
Sn: ≤1 ppb
Sr: ≤0.5 ppb
Tb: ≤0.1 ppb
Te: ≤0.1 ppb
Th: ≤0.1 ppb
Ti: ≤1 ppb
Tl: ≤0.1 ppb
Tm: ≤0.1 ppb
U: ≤0.1 ppb
V: ≤0.5 ppb
W: ≤0.5 ppb
Y: ≤0.1 ppb
Yb: ≤0.1 ppb
Zn: ≤1 ppb
Zr: ≤0.5 ppb

SMILES string

OS(O)(=O)=O

shipped in

ambient

storage temp.

room temp

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

OmniTrace acids offer an economical alternative for sensitive, trace metal analysis and sample preparation. These acids are tested to PPB (parts per billion) levels

Packaging

Product is packed into HDPE bottles. Each HDPE bottle is packed into a single bottle shipping case.

Legal Information

OMNITRACE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service