Documents
SPKGENELC044
Millipore
Spare part, filter mat
W × H × D 89 mm × 89 mm × 10 mm
About This Item
UNSPSC Code:
23151806
material
fibrous
Quality Level
product line
Mobius®
W × H × D
89 mm × 89 mm × 10 mm
compatibility
for use with Mobius®
General description
These filters are used on BioProcessing systems to capture dust, dirt, and other airborne particles. They ensure optimal performance and of the equipment.
This spare part is required for the preventive maintenance service of some of our BioProcessing systems, it is recommended to have one in stock and to change it every 12 months.
Packaging
Pack of 5 units
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany