Skip to Content
Merck

Skip To

SNAPABTR

SNAP id® 2.0 Antibody Collection Tray

Snap i.d. 2.0 Antibody Collection Tray.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116010
NACRES:
NB.22
eCl@ss:
42029053
Technique(s):
western blot: suitable
Species reactivity:
-
Application:
WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

manufacturer/tradename

SNAP id®

technique(s)

western blot: suitable

compatibility

for use with Commercially available blocking reagents, for use with Luminata Western HRP Substrates, for use with Nitrocellulose, for use with PVDF (Immobilon membranes), for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01), for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01), for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001), for use with commercially available detection reagents

detection method

chemiluminescent, colorimetric, fluorometric

shipped in

ambient

storage temp.

room temp

General description

Recover greater than 80% of your Antibody with the Antibody Collection Tray specially designed for the SNAP i.d. systems

Application

For use on immunoblots.

Legal Information

SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SNAP2BHMB050SNAP2MIDISNAP2MB1
technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

western blot: suitable

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

manufacturer/tradename

SNAP id®

manufacturer/tradename

SNAP id®

manufacturer/tradename

SNAP id®

manufacturer/tradename

SNAP id®

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

-

detection method

chemiluminescent, fluorometric, colorimetric

detection method

chemiluminescent, colorimetric, fluorometric

detection method

chemiluminescent, colorimetric, fluorometric

detection method

chemiluminescent, colorimetric, fluorometric




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters