Skip to Content
Merck
All Photos(1)

Key Documents

SLC0NTAIN

Millipore

Millex Product Container

This container is designed for convenient, benchtop storage of 33 mm Millex syringe filters.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24
Pricing and availability is not currently available.

Quality Level

sterility

non-sterile

product line

Millex

manufacturer/tradename

Millex

technique(s)

HPLC: suitable
analytical sample preparation: suitable

shipped in

ambient

General description

Millex Product Container comes with 5 product label cards.The product label cards have space to enter both the catalog and lot number for easy identification of the stored filters. This container can hold up to 200 non-sterile 33 mm Millex syringe filters.

Application

This container is designed for convenient, benchtop storage of 33 mm Millex syringe filters.

Packaging

non sterile

Specifications

For general laboratory use only

Legal Information

Millex is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service