Skip to Content
Merck

Skip To

SIM0000UV

Photooxidation UV Lamp

Photooxidizes organic contaminants. For Simplicity® and Synergy® systems.

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41104200
NACRES:
JA.14
eCl@ss:
33050190


material

stainless steel housing

feature

6 Watts UV lamp

packaging

pkg of 1 unit

parameter

1.0 L/min flow rate (1.2 L/min), 11 kg operating weight

W × D × H

24.8 cm × 26 cm × 46 cm

compatibility

for use with Simplicity® UV, for use with Simplicity®, for use with Synergy® (2001-2006), for use with Synergy® UV

shipped in

ambient

General description

Photooxidation UV Lamp is used to oxidize organic compounds in the feed water.

Application

Photooxidation UV Lamp can be used with Simplicity® and Synergy® water purification systems.

Features and Benefits

  • Genuine consumable for the replacement of SIM0000UV - Photooxidation UV Lamp.
  • Operates at a wavelength of 185 nm

Legal Information

SIMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
SYNERGY is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Photooxidation UV Lamp




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–7 of 7 Questions  
  1. What are the storage conditions for this product?

    1 answer
    1. Recommended storage conditions: temperatures from 5 to 30°C; humidity between 10 and 80%. The original sealed bag (if provided) must be kept intact before the first use to protect the purification element from foreign matter.

      Helpful?

  2. How can I know which consumables can be used with my system?

    1 answer
    1. To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.

      Helpful?

  3. What steps are you taking towards improved sustainability?

    1 answer
    1. Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.

      Helpful?

  4. Can I arrange to have my consumables automatically ordered and delivered once a year?

    1 answer
    1. It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).

      Helpful?

  5. Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?

    1 answer
    1. Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.

      Helpful?

  6. How frequently do the various consumables need to be changed?

    1 answer
    1. The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.

      Helpful?

  7. Can I change the UV lamp myself?

    1 answer
    1. No. With the exception of our SMART system range, a field service engineer must change the UV lamp as this process can be dangerous for customers and requires special protective equipment.

      Helpful?

Reviews

No rating value

Active Filters