Skip to Content
Merck
All Photos(1)

Key Documents

SCT077

Sigma-Aldrich

BioTracker TiNIR HMOX2 Live Cell Dye

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€377.00

€377.00


Estimated to ship on14 May 2025



Select a Size

Change View
1 MG
€377.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

€377.00


Estimated to ship on14 May 2025


Assay

≥98% (HPLC)

form

lyophilized

mol wt

calculated mol wt 797.82

packaging

pkg of 1 mg

manufacturer/tradename

Millipore

storage condition

desiccated
protect from light

technique(s)

cell based assay: suitable

color

green

shipped in

ambient

storage temp.

−20°C

General description

Increasing evidence suggests that a small population of tumor initiating cells (TIC) or cancer stem cells (CSC) are responsible for tumorigenesis and resistance to conventional anti-cancer therapies. Heme oxygenase 2 (HMOX2) has recently been identified as a novel biomarker for tumor initiating cells[1]. BioTracker TiNIR HMOX2 Live Cell Dye is a near-infrared (NIR) fluorescent probe for the detection and isolation of HMOX2 expressing tumor initiating cells. At low concentrations or a single injection of high concentration, TiNIR (tumor initiating cell probe with near infrared) is useful for the isolation of TIC in vitro and in vivo visualization in tumor model animals, respectively. At high concentrations or repeated treatments, TiNIR has been shown to inhibit the function of HMOX2 and induce tumor cell death by accumulating ROS in cancer cells [1].

Spectral Properties
Absorbance: 805 nm
Emission: 825 nm

Reference:1. Kim, J. J., Lee, Y. A., Su, D., Lee, J., Park, S. J., Kim, B., ... & Chang, Y. T. (2019). A near-infrared probe tracks and treats lung tumor initiating cells by targeting HMOX2. Journal of the American Chemical Society, 141(37), 14673-14686.

Application

Purity ≥ 98% confirmed by HPLC. Identification confirmed by HNMR, LC-MS, and elemental analysis.

Features and Benefits

Near-Infrared probe for detection and isolation of HMOX2 expressing tumor initiating cells.

Physical form

Lyophilized. Green solid.

Storage and Stability

Store at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening

Other Notes

Live cell fluorescent imaging

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong-Jin Kim et al.
Journal of the American Chemical Society, 141(37), 14673-14686 (2019-08-23)
Tumor initiating cells (TIC) are resistant to conventional anticancer therapy and associated with metastasis and relapse in cancer. Although various TIC markers and their antibodies have been proposed, it is limited to the use of antibodies for in vivo imaging

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service