Skip to Content
Merck
All Photos(1)

Key Documents

SCT069

Sigma-Aldrich

BioTracker ATP-LW Live Cell Dye

Live cell imaging dye for simultaneous visualization of cellular Adenosine-5′-Triphosphate (ATP) and peroxynitrite (ONOO-).

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€342.00

€342.00


Estimated to ship on20 May 2025



Select a Size

Change View
1 MG
€342.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

€342.00


Estimated to ship on20 May 2025


form

lyophilized

application(s)

cell analysis

detection method

fluorometric

General description

Adenosine-5′-triphosphate (ATP) plays an vital role in regulating cell movement, neurotransmission, and ion channels. Disruption of ATP homeostasis can induce diseases including ischemia, Parkinson′s disease and hypoglycaemia.Peroxynitrite (ONOO-) is a highly reactive nitrogen species with an extremely short biological half-life. ONOO- is known for its deleterious effects, causing irreversible damage to a range of biological targets such as lipids, proteins and nucleic acids. As a result, ONOO- is associated with diseases such as inflammation, cancer, atherosclerosis, ischemia-reperfusion injury and neurodegenerative diseases.BioTracker ATP-LW live cell dye is a dual probe for detection and imaging of ATP and peroxynitrite in live cells. The dye did not respond to presence of other reactive nitrogen/oxygen species such as H2O2 and HClO. However, ADP (adenosine diphosphate) and CTP (cytosine-5′-triphosphate) induced fluorescence enhancements. Note: BioTracker ATP-LW live cell dye is stable in DMSO but unstable is water and aqueous solution. Prepare the working solution and use it immediately. Spectral PropertiesFor ATP detection: Excitation: 450 nmEmission: 550 nmFor Peroxynitrite detection: Excitation: 520 nmEmission: 586 nmResearch Category:InflammationLive Cell Dye

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can you provide additional instructions on the use of it for live cell imaging? This information is not available on the product web page or in the only reference listed for this product (Wu L et al. Dual-function fluorescent probe for the detection of peroxynitrite and adenosine triphosphate. ChemRvix, 27 Feb 2019).

    1 answer
    1. Protocol for Imaging of ATP-LW Staining

      Preparing ATP-LW dye stock solution

      1. Dissolve the contents of one vial (1 mg) of ATP-LW (Molecular Weight: 833.82) in 120 µl of DMSO to create a 10mM solution. Store the stock solution at ≤–20°C for longer periods.

      Labeling cells

      1. Culture cells in an appropriate medium and vessel for fluorescence microscopy.
      2. Dilute the ATP-LW stock solution 1:1,000 ~ 1:50,000 in culture medium to prepare the ATP-LW staining solution.
      3. Remove the medium.
      4. Add sufficient staining solution to cover the cells.
      5. Incubate for 30 minutes, protected from light. Change to culture medium.
      6. Image the cells.

      Spectral information and storage for ATP-LW

      For ATP detection: Excitation/Emission (nm) 450nm/550nm
      For Peroxynitrite detection: Excitation/Emission (nm) 520nm/586nm
      Storage conditions: 2–8°C or ≤–20°C

      Summary of key steps and results:

      - HeLa cells were incubated with Camptothecin for 2h, then ATP-LW probe (30μM) for 30 min.
      - ATP-LW probe works at 15µM~30µM for 30min staining on live Hela cells.
      - Cells preincubated with ATP-LW probe (30μM) exhibited bright fluorescence after treatment with the ONOO− generator 3-morpholinosydnonimine hydrochloride (SIN-1).
      - Green fluorescence increased after treatment with Camptothecin in Hela Cells.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service