Skip to Content
Merck
All Photos(1)

Key Documents

SCT065

Sigma-Aldrich

Green-AlDeSense ALDH1A1 Live Cell Dye

ALDH1A1 isoform specific live cell dye which labels cancer stem cells for fluorescent microscopy, flow cytometry and in vivo applications.

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€562.00

€562.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
€562.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

€562.00


Estimated to ship on30 May 2025


Request a Bulk Order

detection method

fluorometric

General description

Cancer stem cells (CSCs) have an increased capacity to activate antiapoptotic/prosurvival pathways as well as overexpressing ATP-binding cassette transporters which act as potent efflux pumps for potential chemotherapeutics. Aldehyde dehydrogenases (ALDH), in particular the ALDH1A1 isoform, is believed to be a reliable marker of CSCs across many cancer types, including prostate, lung, breast, esophageal, and ovarian cancers. ALDH1A1 is associated with increased chemotherapy resistance and poor clinical outcome and is an established stem cell marker.

Current methods to identify ALDH in living cells are limited by cross-reactivity with several ALDH isoforms producing difficulties in data analysis and prognosis. Chan, et al. have created a ALDH1A1 isoform specific live cell dye termed Green-AlDeSense which labels cancer stem cells and exhibits a 20-fold fluorescent enhancement when bound to ALDH1A1. The dye identifies ALDH1A1 positive CSCs using live cell imaging, flow cytometry and in vivo studies.

Spectral Properties: absmax: 496 nm, emmax: 516 nm.

Storage and Stability

Store at -20C. Protect from light.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service