Skip to Content
Merck
All Photos(1)

Documents

SCT056

Sigma-Aldrich

BioTracker QUMA-1 RNA G-quadruplex Live Cell Dye

A red-emitting fluorescent probe for tracking RNA G-quadruplexes dynamics, such as folding and unfolding, in live cells.

Synonym(s):

Live cell imaging probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

Assay

≥98% (H-NMR)
≥98% (HPLC)
≥98% (LC/MS)
≥98% (elemental analysis)

technique(s)

cell based assay: suitable

detection method

fluorometric

General description

RNA G-quadruplexes are formed by self-assembly of guanine-rich RNA stacks into stacked G-quartets. This can occur in protein-coding (mRNA) as well as non-coding (ncRNA) transcripts. There is growing evidence that RNA G-quadruplexes are involved in different biological functions: telomere elongation, DNA recombination, transcription, RNA post-transcriptional mechanisms (pre-mRNA processing, mRNA turnover, targeting and translation), and are associated with sevral human diseases.

BioTracker<TMSYMBOL></TMSYMBOL> QUMA-1 RNA G-quadruplex Live Cell Dye (QUMA-1) is a red-emitting fluorescent probe that can be used to study dynamics (folding and unfolding) and movements of RNA G-quadruplexes. QUMA-1 is a reversible dye that emits upon binding to G-quadruplexes and turns off upon unfolding. QUMA-1 is highly specific for RNA G-quadruplexes and does not bind to DNA, double-stranded RNA, single-stranded RNA or other RNA structures. QUMA-1 could also potentially be used to study RNA-protein interactions.

Spectral Properties
Absorbance: 555 nm
Emission: 660 nm

Application

A red-emitting fluorescent probe for tracking RNA G-quadruplexes dynamics, such as folding and unfolding, in live cells.
Live cell fluorescent imaging
Research Category
Cell Imaging

microRNA

RNA Metabolism & Binding Proteins
Research Sub Category
Live Cell Dye

Quality

Purity: ≥ 98% confirmed by HNMR, LC-MS and HPLC and elemental analysis
Molar Mass: 628.17 g/mol

Physical form

Lyophilized

Storage and Stability

Store BioTracker QUMA-1 RNA G-quadruplex -Live Cell Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiu-Cai Chen et al.
Angewandte Chemie (International ed. in English), 57(17), 4702-4706 (2018-02-18)
Because of the absence of methods for tracking RNA G-quadruplex dynamics, especially the folding and unfolding of this attractive structure in live cells, understanding of the biological roles of RNA G-quadruplexes is so far limited. Herein, we report a new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service