SCM112
3dGRO® Wnt3a Conditioned Media Supplement
Synonym(s):
Wnt3a Conditioned Media;Wnt3a CM;Wnt Conditioned Media
About This Item
Recommended Products
packaging
1 ea of 50 mL
manufacturer/tradename
Millipore
technique(s)
cell culture | mammalian: suitable
cell culture | stem cell: suitable
shipped in
dry ice
storage temp.
−20°C
Cell Line Description
Application
- Appearance (color): clear, red liquid
- Endotoxin: 2 EU/mL
- Sterility Tested: No Growth/Pass
- Mycoplasma: Negative
- Media tested for function in supporting growth of human pancreatic organoids
- Media tested for biological activity using a Wnt3a luciferase reporter assay in HEK293 STF cells
Features and Benefits
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service