Skip to Content
Merck

SCC448

OSUMMER.4 Mouse NRAS-Mutant Melanoma Cell Line

Mouse

Synonym(s):

Mouse melanoma cell line

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Biological source:
mouse
Growth mode:
N/A
Manufacturer/tradename:
Millipore
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

OSUMMER.4 Mouse NRAS-Mutant Melanoma Cell Line,

biological source

mouse

Quality Segment

packaging

vial of ≥1X10⁶ cells/vial vial

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

Application

Each vial contains >1X106 viable cells.

Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel and have been found to be negative for all pathologies in this test.

Cells are verified to be of mouse origin and negative for interspecies contamination from rat, human, Chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination Clear panel.

Cells are negative for mycoplasma contamination.

Cells are genotyped by STR analysis to verify the unique identity of the cell line.
The OSUMMER (Ohio State University and Moffitt Melanoma Exposed to Radiation) cell lines fulfill a previously unfilled gap in melanoma biology. These cell lines are syngeneic with C57BL/6 laboratory mice and have genetic profiles that are similar to those of human tumors, making them responsive to immunotherapy treatments. These NRAS-mutant cell lines can be used to discover and address potential immunotherapies in NRAS-mutant human melanomas, which make up an estimated 15-30% of human melanomas. These cell lines also enable in vivo testing of immunotherapies with a mouse model.

OSUMMER.6 NRAS mouse melanoma cell line was derived from a male TN mouse (homozygous for the LSL-NrasQ61R allele) exposed to UVA and UVB radiation. OSUMMER.4 can form tumors in vivo without Matrigel injection. WES (whole exome sequencing) revealed high SNV (Single Nucleotide variants) and low CNA (copy number alterations) burden, as well as the presence of UV signature mutations. OSUMMER.4 cells express the melanoma markers PMEL and MelanA and exhibit robust growth in culture. OSUMMER.6 cells demonstrate a relatively low (<lt/>200) mutation burden compared with other OSUMMER NRAS Q61R mutants, and are resistant to MEK inhibition (MEKi).

Source
OSUMMER.6 was derived from melanoma induced in TN (Tyr::CreER) mice exposed to UVA or UVB radiation on postnatal day 3 (1).

References
1. Bowman, R. L., et al. (2021). UVB mutagenesis differs in Nras-and Braf-mutant mouse models of melanoma. Life Science Alliance, 4(9).

Features and Benefits

The OSUMMER.4 melanoma cell line carries the NRAS muation Q61R and may be used to create syngeneic mouse melanoma models.

Preparation Note

Cells should be stored in liquid nitrogen. Cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact [email protected].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library