Skip to Content
Merck

Skip To

SCC322

3dGRO® Duodenum Intestinal Organoids, Age 21 (Prep 87-D)

Normal, Duodenum, Age: 21, Male, Mutation: None

Synonym(s):

3D organoid culture, Intestinal organoids

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.81
UNSPSC Code:
41106509
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human (duodenum)

description

Cryopreserved patient derived gastrointestinal organoids (PDOs)

packaging

vial of >1500 viable organoids

gender

male

technique(s)

cell culture: suitable

application(s)

3D cell culture

control

(duodenum)

relevant disease(s)

(Normal)

storage temp.

−196°C

General description

Patient derived organoids (PDOs) are novel in vitro 3D cell models that preserve original tissue physiology and molecular pathology, thus representing a clinically-relevant alternative to traditional 2D cell lines and an effective tool to refine and reduce animal models. PDOs can be derived from adult patient biopsies or resected tissues containing native LGR5+ stem cell populations and cultured within an ECM-rich substrate using specialized organoid media such as L-WRN conditioned media. Adult tissue derived organoids are phenotypically and genetically stable in long term culture, presenting more mature phenotypes compared to iPSC-derived organoids. Importantly, PDOs have shown to be able to predict patient clinical responses to chemotherapeutics.
We are now offering a comprehensive biobank of highly characterized tissue-derived human gastrointestinal organoids from normal and diseased patients. The intestinal organoid biobank contains over 50 highly characterized intestinal organoids from both normal and diseased patients derived from multiple regions of the digestive systems including small intestine (duodenum, ileum), stomach, rectum and colon.
Cell Line Characteristics
  • Sex: Male
  • Age: 21 Years
  • Organ: Duodenum
  • Disease: Normal

Packaging

≥1500 viable organoids/vial

Preparation Note

Store in liquid nitrogen. The organoids can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Legal Information

3dGRO® is a trademark of Merck KGaA, Darmstadt, Germany.
3dGRO® organoids were derived utilizing HUB Organoid Technology.

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/pdouse. This product may not be re-engineered or copied; used to make derivatives or modifications; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact [email protected].
3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCC321SCC324SCC316
technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

biological source

human (duodenum)

biological source

human colon

biological source

human (duodenum)

biological source

human (duodenum)

relevant disease(s)

(Normal)

relevant disease(s)

(Normal)

relevant disease(s)

(Normal)

relevant disease(s)

(Normal)

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

packaging

vial of >1500 viable organoids

packaging

vial of >1500 viable organoids

packaging

vial of >1500 viable organoids

packaging

vial of >1500 viable organoids


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters