Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 vial | Check Cart for Availability | € 2.590,00 |
About This Item
€ 2.590,00
biological source
human (duodenum)
description
Cryopreserved patient derived gastrointestinal organoids (PDOs)
packaging
vial of >1500 viable organoids
gender
female
technique(s)
cell culture: suitable
application(s)
3D cell culture
control
(duodenum)
relevant disease(s)
(Normal)
storage temp.
−196°C
General description
We are now offering a comprehensive biobank of highly characterized tissue-derived human gastrointestinal organoids from normal and diseased patients. The intestinal organoid biobank contains over 50 highly characterized intestinal organoids from both normal and diseased patients derived from multiple regions of the digestive systems including small intestine (duodenum, ileum), stomach, rectum and colon.
Cell Line Characteristics
- Sex: Female
- Age: 62 Years
- Organ: Duodenum
- Disease: Normal
Packaging
Preparation Note
Legal Information
3dGRO® organoids were derived utilizing HUB Organoid Technology.
Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/pdouse. This product may not be re-engineered or copied; used to make derivatives or modifications; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact [email protected].
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| application(s) 3D cell culture | application(s) 3D cell culture | application(s) 3D cell culture | application(s) 3D cell culture |
| technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable |
| relevant disease(s) (Normal) | relevant disease(s) (Normal) | relevant disease(s) (Normal) | relevant disease(s) (Normal) |
| biological source human (duodenum) | biological source human (duodenum) | biological source human (duodenum) | biological source human colon |
| storage temp. −196°C | storage temp. −196°C | storage temp. −196°C | storage temp. −196°C |
| packaging vial of >1500 viable organoids | packaging vial of >1500 viable organoids | packaging vial of >1500 viable organoids | packaging vial of >1500 viable organoids |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



