SCC317
3dGRO® Colon Intestinal Organoids, Age 56 (Prep 84-C)
Cryopreserved patient derived gastrointestinal organoids (PDOs)
Select a Size
€2,490.00
Estimated to ship on29 May 2025
Select a Size
About This Item
€2,490.00
Estimated to ship on29 May 2025
Recommended Products
technique(s)
cell culture | stem cell: suitable
General description
We are now offering a comprehensive biobank of highly characterized tissue-derived human gastrointestinal organoids from normal and diseased patients. The intestinal organoid biobank contains over 50 highly characterized intestinal organoids from both normal and diseased patients derived from multiple regions of the digestive systems including small intestine (duodenum, ileum), stomach, rectum and colon.
Cell Line Characteristics
- Sex: Female
- Age: 56 Years
- Organ: Colon
- Disease: Normal
Cell Line Origin
Packaging
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service