Skip to Content
Merck
All Photos(1)

Key Documents

SCC256

Sigma-Aldrich

BPH-1 Human Benign Prostatic Hyperplasia Cell Line

The BPH-1 cell line is a well-established model for human prostate biology and widely used for investigation into the causes and potential treatments for prostatic disease.

Synonym(s):

BPH-1 cells, Benign Prostatic Hyperplasia cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€1,010.00

€1,010.00


Estimated to ship on24 May 2025



Select a Size

Change View
1 VIAL
€1,010.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

€1,010.00


Estimated to ship on24 May 2025


application(s)

cell analysis

General description

BPH-1 is an immortalized benign prostatic hyperplasia cell line that is a highly cited human model for prostate growth and physiology. Compared to prostate cancer cell lines, BPH-1 cells expressed increased levels of estradiol (E2) and aromatase, factors that have been implicated in BPH progression (2). BPH-1 cells share the cytokeratin profile of luminal epithelial cells and do not express androgen receptor or prostate-specific antigen (PSA) (3). BPH-1 cells express the proto-oncogene p53 and harbor several chromosomal structural rearrangements but are non-tumorigenic in nude mice (3). Source:The BPH-1 cell line was established from primary prostatic tissue from a 68-year-old patient and the cell line was immortalized with Simian virus 40 (SV40) large T antigen (3). BPH-1 cells contain a neomycin resistance cassette and are resistant to G418.

Cell Line Origin

Human, Epithelial Cells

Packaging

≥1X106 cells/vial

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Disclaimer

This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service