Skip to Content
Merck
All Photos(4)

Key Documents

SCC127M

Sigma-Aldrich

SF8628 Human DIPG H3.3-K27M Cell Line

Human

Synonym(s):

SF-8628 cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€1,230.00

€1,230.00


Estimated to ship on05 April 2025



Select a Size

Change View
1 VIAL
€1,230.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

€1,230.00


Estimated to ship on05 April 2025


Product Name

SF8628 Human DIPG H3.3-K27M Cell Line, SF8628 pediatric diffuse intrinsic pontine glioma (DIPG) cell line harbors the histone H3.3 Lys 27-to-methionine (K27M) mutation and can support research and drug development efforts targeting DIPG.

biological source

human

Quality Level

technique(s)

cell culture | mammalian: suitable

General description

Diffuse intrinsic pontine gliomas (DIPG) are highly aggressive and difficult to treat tumors arising in the ventral pons of the brain stem. Despite therapeutic advances, DIPG is incurable and most patients, primarily children, die within 2 years of diagnosis. DIPG is one of the leading causes of death in children with brain tumors (1).

A somatic mutation of histone H3.3 resulting in a lysine 27 to methionine substitution (H3.3K27M) occurs in 60% of DIPG (2). In H3.3K27M DIPG patient samples, levels of H3K27 dimethylation (H3K27me2) and trimethylation (H3K27me3) are reduced globally. These epigenetic changes are thought to be important factors driving DIPG oncogenesis (2,3,4). Expression of H3.3K27M was also recently shown to be associated with increased levels of H3K27 acetylation (H3K27ac) and recruitment of bromodomain proteins at sites of active transcription (5). Treatment of H3.3K27M DIPG cells with a bromodomain and extra-terminal domain (BET) inhibitor was found to inhibit proliferation and induced differentiation. BET inhibitors are thus a promising therapeutic approach for the treatment of DIPG.

1. Schroeder KM et al. (2014) Pediatr. Res. 75(1-2): 205–209.
2. Lewis PW et al. (2013) Science 340(6134): 857–861.
3. Chan K-M et al. (2013) Genes & Development 27: 985-990.
4. Hashizume R et al. (2014) Nat Med. 20(12):1394-6.
5. Piunti A et al. (2017) Nat. Med. 23(4): 493-500.
6. Mueller S et al. (2014) Neuro Oncol. 16(3): 352-360.
SF8628 is a human glioma cell line derived by surgical biopsy from a pediatric H3.3K27M DIPG patient (6). This novel cell line may be used to facilitate the testing of specific targeted therapeutics for DIPG.

Cell Line Description

Cancer Cells

Application

Research Category
Cancer

Oncology
SF8628 pediatric diffuse intrinsic pontine glioma (DIPG) cell line harbors the histone H3.3 Lys 27-to-methionine (K27M) mutation and can support research and drug development efforts targeting DIPG.
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested negative for Epstein-Barr virus, HPV-16, HPV-18, Hepatitis A, C, Herpesvirus type 6, 7, 8 and HIV-1 & 2 viruses by PCR
• Cells are negative for mycoplasma contamination.
• Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. En qué medio de cultivos crece la línea celular SF8628

    1 answer
    1. The recommended medium for this cell line is as follows:
      DMEM-High Glucose - D6546
      10 % FBS - ES-009-B
      2 mM L-Glutamine - TMS-002-C
      1X Penicillin-Streptomycin Solution - TMS-AB2-C

      Please see the links below to review the product datasheet:
      https://www.emdmillipore.com/US/en/product/SF8628-Human-DIPG-H3.3-K27M-Cell-Line,MM_NF-SCC127#anchor_DS

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service