SCC067
HMC-1.1 Human Mast Cell Line
Human
Synonym(s):
HMC-1 cell line
About This Item
Recommended Products
Product Name
HMC-1.1 Human Mast Cell Line, HMC-1.1 human mast cell line is a variant subline of the HMC-1 cell line and possesses the V560G KIT mutation.
biological source
human
Quality Level
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
General description
1. Nilsson, G, et al. (1994) Phenotypic Characterization of the Human Mast-Cell Line HMC-1. Scand. J. Immunol 39: 489-498.
2. Sundstrom, M, et al. (2003) Functional and phenotypic studies of two variants of a human mast cell line with a distinct set of mutations in the c-kit proto-oncogene. Immunology 108: 89-97.
3. Furitsu, T, et al. (1993) Identification of mutations in the coding sequence of the proto-oncogene c-kit in a human mast cell leukemia cell line causing ligand-independent activation of c-kit product. J. Clin. Invest. 92(4): 1736-4.
Cell Line Description
Application
Inflammation & Immunology
Cancer
Quality
• Cells are tested by PCR and are negative for HPV-16, HPV-18, Hepatitis A, B, C and HIV-1 & 2 viruses.
• Cells are negative for mycoplasma contamination.
• Each lot of cells are genotyped by STR analysis to verify the unique identity of the cell line.
Storage and Stability
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service