Skip to Content
Merck
All Photos(1)

Key Documents

S22-M

Sigma-Aldrich

Sheep Serum, 100 ml

Non-sterile sheep serum collected from normal healthy sheep. Used for blocking of interfering antibodies.

Synonym(s):

100 ml Serum

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€235.00

€235.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€235.00

About This Item

UNSPSC Code:
41105901
eCl@ss:
26020100
NACRES:
NA.71
biological source:
sheep

€235.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

sheep

antibody form

serum

manufacturer/tradename

Chemicon®

technique(s)

blocking: suitable (interfering antibodies)

General description

Normal sheep serum collected from healthy normal sheep.

Physical form

Nonsterile liquid. Contains no preservatives. Filtered through a .45 micron membrane filter.

Storage and Stability

Store frozen at -20°C. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service