Skip to Content
Merck
All Photos(1)

Key Documents

S20-M

Sigma-Aldrich

Rabbit Serum, 100 ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105901
eCl@ss:
26020100
NACRES:
NA.71
biological source:
rabbit
Pricing and availability is not currently available.

biological source

rabbit

antibody form

serum

manufacturer/tradename

Chemicon®

technique(s)

blocking: suitable (interfering antibodies)

General description

Normal rabbit serum collected from healthy normal rabbits.



Serum Preparation: Blood is collected from New Sealand White or California White rabbits and allowed to clot at room temperature. The preparation is rimmed and then centrifuged to separate the clotted material from the serum. Supernatant is retained and pooled.



Characterisitcs/Measurements Recorded:

Appearance - Yellow to red liquid

pH - 7.5 to 7.9

Total Protein - 5.5 to 6.5 gm

Hemoglobin - 20 to 25 mg/dL

Physical form

Nonsterile liquid. Contains no preservatives. Filtered through a .45 micron membrane filter.

Storage and Stability

Store frozen at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael G Wade et al.
Biology of reproduction, 78(5), 822-831 (2008-01-18)
We used high-density microarrays to evaluate the possible mechanisms by which 2-methoxyacetic acid (MAA) disrupts spermatogenesis. Levels of mRNA transcripts were determined in total RNA isolated from testes of MAA-treated (650 mg/kg i.p.) or concurrent control rats killed 4, 8

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service