Skip to Content
Merck

Skip To

PSET010

Millicell® 24 well Plates

pore size 8.0 μm, polycarbonate membrane, sterile, Tissue Culture (TC)-treated surface, automation compatible

Synonym(s):

24-Well Plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 179,00
5 ea
Check Cart for Availability
€ 740,00

About This Item

UNSPSC Code:
41104923
NACRES:
NB.22
eCl@ss:
32039006
Material:
polycarbonate membrane, polystyrene plate
Pore size:
8.0 μm
Sterility:
sterile
Binding type:
Tissue Culture (TC)-treated surface

€ 179,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

polycarbonate membrane, polystyrene plate

description

  • 1/pack components: 24-well cell culture plate (1), single well feeder tray (1), 24-well receiver plate (1), lid (2)
  • 5/pack components: 24-well cell culture plates (5), 24 well receiver plate (5), lid (5)

sterility

sterile

feature

lid, automation compatible

manufacturer/tradename

Millicell®

parameter

50 °C max. temp.

technique(s)

cell attachment: suitable, cell culture | mammalian: suitable, cell differentiation: suitable, cell migration: suitable

L × W × H

127.76 mm × 85.48 mm × 20.32 mm

filter diam.

9.5 mm

filtration area

0.7 cm2

plate size

24 wells

working volume

0.4 mL

matrix

Isopore

pore size

8.0 μm

binding type

Tissue Culture (TC)-treated surface

shipped in

ambient

storage temp.

10-30°C

General description

Millicell cell culture insert plates are a patented device designed to support cell growth and differentiation of endothelial and epithelial cell lines, including CaCo-2.

The Millicell device is available with a 0.4 µm PCF and a variety of PET membrane pore sizes. The plate is designed for maximum user convenience and incorporates features such as apical assist for easier pipetting and basolateral access. To reduce the risk of monolayer contamination, the membrane plates have feet to elevate the plate above the work surface when disassembled from the feeder tray. Tear drop-shaped receiver wells eliminate air bubbles as plates are assembled.

Millicell plate are optimized to grow and sustain high integrity cell monolayers. Cells grown on Millicell plate membranes grow better than on plastic because the cells are nourished from both the apical and basolateral sides. Cell growth and function more closely mimic in vivo. Millicell plates are designed for analysis as well as cell growth and can be used manually or with automated cell seeding, feeding and washing systems

With this complete system, you can grow, feed, and analyze cells in one membrane-bottom plate. The plate can be used with either a single-well or a multi-well feeding tray. At the time of transport analysis, the plate is simply transferred to a multi-well transport tray for analysis.

This streamlined design enhances functionality with:

- Seed-and-feed systems

- Automated liquid handling systems and span pipettes

- Basolateral and apical access to cells

- Transepithelial electrical resistance (TEER) measurement systems

Legal Information

ISOPORE is a trademark of Merck KGaA, Darmstadt, Germany
Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PSHT010RPSST010RPSMT010R
material

polycarbonate membrane, polystyrene plate

material

polycarbonate membrane, polystyrene plate

material

polycarbonate membrane, polystyrene

material

polycarbonate membrane, polystyrene plate

sterility

sterile

sterility

sterile

sterility

γ-irradiated, sterile

sterility

γ-irradiated, sterile

manufacturer/tradename

Millicell®

manufacturer/tradename

Millicell®

manufacturer/tradename

Millicell®

manufacturer/tradename

Millicell®

binding type

Tissue Culture (TC)-treated surface

binding type

Tissue Culture (TC)-treated surface

binding type

Tissue Culture (TC)-treated surface

binding type

Tissue Culture (TC)-treated surface

pore size

8.0 μm

pore size

0.4 μm

pore size

3 μm

pore size

5 μm

feature

lid, automation compatible

feature

lid, 100x106 pores/cm2 (pore density), automation compatible

feature

lid, automation compatible

feature

lid, automation compatible


Still not finding the right product?

Explore all of our products under Millicell® 24 well Plates


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters