Skip to Content
Merck
All Photos(1)

Key Documents

PS1019

Sigma-Aldrich

PhosphoDetect Anti-JNK1/2 (pThr¹⁸³/Tyr¹⁸⁵) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-c-Jun N-terminal Kinase 1&2, Phospho-Specific (Thr/Tyr 183/185), Anti-c-Jun N-terminal Kinase 1&2, Phospho-Specific (Thr/Tyr183/185)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€560.00

€560.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€560.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€560.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr183/pTyr185)

Gene Information

human ... MAPK8(5599)

General description

Rabbit polyclonal antibody adsorbed against non-phosphopeptide corresponding to the immunogen phosphorylation site, followed by immunoaffinity chromatography. Recognizes the ~49 kDa JNK1 and the ~55 kDa JNK2 proteins phosphorylated at Thr183 and Tyr185.
Recognizes the ~49 kDa JNK1 and the ~55 kDa JNK2 phosphorylated at Thr183 and Tyr185 in uv-treated HEK293 cells.
This PhosphoDetect Anti-JNK1/2 (pThr¹⁸³/Tyr¹⁸⁵) Rabbit pAb is validated for use in Immunoblotting for the detection of JNK1/2 (pThr¹⁸³/Tyr¹⁸⁵).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Thr¹⁸³ and Tyr¹⁸⁵ phosphorylation sites of human JNK1 and JNK2

Application


Immunoblotting (1:1000)

Warning

Toxicity: Standard Handling (A)

Physical form

In Dulbecco′s PBS (without Mg2+ and Ca2+), 1 mg/ml BSA, 50% glycerol, pH 7.2-7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
UV-treated HEK293 cells or sorbitol-treated PC12 cells

Other Notes

Choudhury, B.K., et al. 2002. J. Immunol.169, 5955.
Barr, R.K., et al. 2001. Int. J. Biochem. Cell. Biol.33, 1047.
Dong, C., et al. 2001. J. Clin. Immunol.21, 253.
Janulis, M., et al. 2001. Mol. Cell. Biol.21, 2235.
Pearce, A.K., et al. 2001. Trends Cell Biol.11, 426.
Tong, T., et al. 2001. Exp. Cell Res.269, 64.
Cobb, M.H., et al. 1999. Prog. Biophys. Mol. Biol.71, 479.
Basu, S., et al. 1998. Oncogene17, 3277.
The immunogen sequence is highly conserved among many species, including mouse, rat, chicken, nematode, and Drosophila, but cross-reactivity has not been determined. The immunogen sequence is also conserved in JNK3, but reactivity has not been detected. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Hoste et al.
Nature communications, 12(1), 5913-5913 (2021-10-10)
OTULIN is a deubiquitinase that specifically cleaves linear ubiquitin chains. Here we demonstrate that the ablation of Otulin selectively in keratinocytes causes inflammatory skin lesions that develop into verrucous carcinomas. Genetic deletion of Tnfr1, knockin expression of kinase-inactive Ripk1 or
Lien Verboom et al.
Cell death & disease, 14(8), 534-534 (2023-08-20)
The intestinal epithelium is a single cell layer that is constantly renewed and acts as a physical barrier that separates intestinal microbiota from underlying tissues. In inflammatory bowel disease (IBD) in humans, as well as in experimental mouse models of
Kyubin Lee et al.
Biology, 9(8) (2020-08-13)
Abeliophyllum distichum Nakai is known as a monotypic genus endemic to South Korea. Currently, several pharmacological studies have revealed that A. distichum extract exhibits diverse biological functions, including anti-cancer, anti-diabetic, anti-hypertensive, and anti-inflammatory activities. In this study, we present the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service