PRCYTA-40K-PX38
MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A 38-Plex Premixed Magnetic Bead Panel
Synonym(s):
38-Plex Cytokine Beads, Cytokine Bead Panel, Primate Cytokine Panel
About This Item
species reactivity
nonhuman primates
assay range
accuracy: 80-102%
inter-assay cv: <20%
intra-assay cv: <10%
inter-assay cv: <30%
(TNF-α)
technique(s)
multiplexing: suitable
detection method
fluorometric (Luminex® xMAP®)
shipped in
wet ice
storage temp.
2-8°C
General description
The MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A 38-Plex Premixed Magnetic Bead Panel is a 38-plex kit to be used for the simultaneous quantification of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: sCD137, CD40L, sFASL, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNα2, IFNγ, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, MIP-3α, Perforin, RANTES, TGFα, TNFα, VEGF-A.
Specificity
Application
- Analytes: sCD137, CD40L, sFASL, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNα2, IFNγ, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, MIP-3α, Perforin, RANTES, TGFα, TNFα, VEGF-A
- Recommended Sample type: Serum, plasma or tissue/cell lysate and culture supernatant samples
- Recommended Sample dilution: Neat plasma or serum. Tissue culture supernatant may require a dilution with an appropriate control medium prior to assay. RANTES requires a 1:100 dilution of plasma/serum samples.
NOTE: RANTES cannot be plexed with other cytokines in this panel due to a required 1:100 dilution of plasma/serum samples. This premixed panel comes with a RANTES bead, so please plan your samples accordingly.
- Note: The serum/plasma samples of some non-human primate species may require up to 1:100 dilution in Assay Buffer for IL-8.
- Assay Run Time: Overnight or two-hour primary incubation. For best results, an overnight incubation is recommended
- Research Category: Inflammation & Immunology
- Research Sub Category: Obesity, Metabolic Disorders, Inflammation & Autoimmune Mechanisms
Features and Benefits
Storage and Stability
- Recommended storage for kit components is 2 - 8°C.
- For long-term storage, freeze reconstituted standards and controls at -20°C. Avoid multiple freeze/thaw cycles.
- DO NOT FREEZE Antibody-Immobilized Beads, Detection Antibody, and Streptavidin-Phycoerythrin.
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
Target Organs
Respiratory Tract
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Multiplex like never before with the MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A that offers researchers the ability to profile the largest selection of immune factors available for non-human primates on one 96-well plate.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service