Skip to Content
Merck
All Photos(1)

Key Documents

PMEF-DR4-M

Sigma-Aldrich

EmbryoMax® Primary Mouse Embryonic Fibroblasts

Mouse

Synonym(s):

MEF Feeder Cells, DR4 PMEFs, DR4 MEFs, MEFs, Mouse Feeder Cells

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€193.00
5 VIALS
€801.00

€193.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
1 VIAL
€193.00
5 VIALS
€801.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

€193.00


Estimated to ship on27 May 2025


Request a Bulk Order

Product Name

EmbryoMax® Primary Mouse Embryonic Fibroblasts, PMEF, Strain DR4, Mitomycin C Treated, Passage 3

biological source

mouse

Quality Level

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | stem cell: suitable

General description

Plating MEF Feeder Cells

Procedure:

1. Prior to thawing PMEF feeder cells, coat plates/flasks with Gelatin solution.
2. Thaw PMEF vial(s) quickly in a 37 °C water bath and transfer to a 15 mL tube (already containing 10 mL of warm PMEF Feeder Cell Medium). Gently invert the tube to distribute, and centrifuge at 300 xg for 4–5 minutes.
3. Remove supernatant and resuspend the cell pellet in warm PMEF Feeder Cell Medium.
4. Remove the Gelatin solution from plates/flasks, and aliquot the PMEF feeder cell suspension at the densities recommended in Table 4.1 of the mouse ES protocol guide
5. Incubate the PMEF Feeder cells at 37 °C with 5% CO2. Use Figures 4A, B and C in the mouse ES protocol guide as a guide for an estimate of correct PMEF density and
appearance. Gelatinized plates may be used for 12–14 days.
The EmbryoMax range of PMEF cells provides researchers with a convenient solution for ES cell culture by eliminating the need for time consuming feeder cell isolation and preparation. Many embryonic stem cell culture protocols necessitate the use of primary mouse embryo fibroblast (PMEF) cells. In these protocols, ES cells are typically cultured on a monolayer of PMEF feeder cells. Feeder cells perform two important roles in stem cell culture: they secrete several important growth factors into the medium, which help maintain pluripotency, and they provide a cellular matrix for ES cells to grow.

The DR4 strain of MEF feeder cells are resistant to neomycin, hygromycin, puromycin, and 6-thioguanine. They are mitotically arrested by mytomycin-C treatment and will not proliferate.

Cell Line Description

Primary Mouse Embryo Fibroblasts

Components

EmbryoMax PMEF, P3, Strain DR4, Mito-C Treated: (Part number PMEF-DR4) One (1) vial containing ≥4 x10^6 cells per vial.

Quality

Cell Viability and Morphology: PASSED
Mycoplasma Testing: PASSED
Bacterial Testing: PASSED
Fungi Testing: PASSED

Storage and Stability

Vials should be stored in the vapor phase of liquid nitrogen.

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service