Skip to Content
Merck

Skip To

PC154

Anti-Excitatory Amino Acid Transporter (GLT-1) Rabbit pAb

liquid, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 583,00

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
rat
Application:
Citations:
1

€ 583,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

diluted serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Immunoblotting (1:1000)
Immunoprecipitation )

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Rattus norvegicus ... Slc1A2(29482)

General description

Anti-Excitatory Amino Acid Transporter (GLT-1), rabbit polyclonal recognizes the ~75 kDa and the ~50 kDa forms of GLT-1. It is validated for use in Western blotting and immunoprecipitation.
Rabbit polyclonal antibody supplied as diluted serum. Recognizes the ~75 kDa and the ~50 kDa forms of GLT-1.
Recognizes the ~75 kDa and the ~50 kDa forms of GLT-1.

Immunogen

a synthetic peptide (NGKSADCSVEEEPWKREK) corresponding to amino acids at the C-terminus of rat GLT-1

Application

Immunoblotting (1:1000)Immunoprecipitation

Other Notes

Arriza, J.L., et al. 1997. Proc. Natl. Acad. Sci. USA94, 4155.
Nagao, S., et al. 1997. Neuroscience78, 929.
Rothstein, J.D., et al. 1995. Ann. Neurol.14, 5559.
Arriza, J.L., et al. 1994. J. Neurosci.14, 5559.
Rothstein, J.D., et al. 1993. Proc. Natl. Acad. Sci. USA90, 6591.
Kanai, Y., et al. 1992. Nature360, 467.
Pines, G., et al. 1992. Nature360, 464.
Rothstein, J.D., et al. 1992. N. Engl J. Med.326, 1464.
Storck T., et al. 1992. Proc. Natl. Acad. Sci. USA89, 10955.
This antibody may also cross react nonspecifically with a ~102 kDa protein not believed to be related to GLT-1. This antibody has also been reported to work for immunoprecipitation. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
178467400062437695
description

liquid, Calbiochem®

description

liquid, Calbiochem®

description

liquid, Calbiochem®

description

liquid, Calbiochem®

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

antibody form

diluted serum

antibody form

diluted serum

antibody form

affinity isolated antibody

antibody form

diluted serum

biological source

rabbit

biological source

goat

biological source

rabbit

biological source

rabbit

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

form

liquid

form

liquid

form

liquid

form

liquid

Gene Information

Rattus norvegicus ... Slc1A2(29482)

Gene Information

-

Gene Information

human ... SLC2A3(6515)

Gene Information

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters