NX0412
Nitric Acid
Synonym(s):
Nitric acid solution, Hydrogen Nitrate, Azotic Acid,Aqua Fortis, Aquafortis
About This Item
Recommended Products
vapor pressure
9.4 hPa ( 20 °C)
Quality Level
concentration
64.8-65.2% HNO3
impurities
≤0.005 ppm Arsenic (As)
≤0.08 ppm Chloride (Cl)
≤0.08 ppm Sulfate (SO4)
≤0.2 ppm Heavy metals (as Pb)
≤0.2 ppm Iron (Fe)
≤0.2 ppm Phosphate (PO4)
≤1 ppm Fluorine (F)
≤5 ppm Nonvolatile matter
pH
<1 (20 °C in H2O)
bp
121 °C/1013 hPa
mp
-32 °C
shipped in
ambient
storage temp.
room temp
SMILES string
O[N+]([O-])=O
InChI
1S/HNO3/c2-1(3)4/h(H,2,3,4)
InChI key
GRYLNZFGIOXLOG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Analysis Note
Assay: 64.8-65.2% by wt.
Chloride (Cl): 0.08ppm max
Fluorine (F): 1ppm max
Heavy metals (as Pb): 0.2ppm max
Iron (Fe): 0.2ppm max
Nonvolatile matter: 5ppm max
Phosphate (PO4): 0.2ppm max
Sulfate (SO4): 0.08ppm max
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A
Supplementary Hazards
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service