Skip to Content
Merck
All Photos(1)

Documents

NX0408

Supelco

Nitric Acid

OmniTrace Ultra

Synonym(s):

Nitric acid solution, Hydrogen Nitrate, Azotic Acid,Aqua Fortis, Aqua Fortis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352301

grade

for inorganic trace analysis

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8

vapor pressure

9.4 hPa ( 20 °C)

form

liquid

concentration

≥65-70% in nitric acid

pH

<1 (20 °C in H2O)

bp

121 °C/1013 hPa

mp

-32 °C

solubility

soluble (Water: Soluble)

density

1.39 g/cm3 at 20 °C (68 °F)

shipped in

ambient

storage temp.

room temp

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

OMNITRACE® Ultra
Product is double-distilled and packed in class 100 clean room conditions into preleached FEP bottles. Each FEP bottle is packed in an EPS innerpack and then packed into a 1x regulated carton.

Legal Information

OMNITRACE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service