Skip to Content
Merck
All Photos(1)

Key Documents

NX0403

Sigma-Aldrich

Ninhydrin

Synonym(s):

Ninhydrin

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
Inquire


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
MDL number:
UNSPSC Code:
12352115
NACRES:
NA.21


Estimated to ship on28 May 2025


Request a Bulk Order

manufacturer/tradename

Calbiochem®

Quality Level

storage condition

OK to freeze

shipped in

ambient

SMILES string

OC1(C(=O)c2c(cccc2)C1=O)O

InChI

1S/C9H6O4/c10-7-5-3-1-2-4-6(5)8(11)9(7,12)13/h1-4,12-13H

InChI key

FEMOMIGRRWSMCU-UHFFFAOYSA-N

General description

Suitable for biochemistry.

Specifications:
Assay 99% min
Identity (IR-spectrum): Conforms
pH (1%, water): 4.6-5.6
Suitability for detection of amino acids: To pass test
Sulfated ash: 0.1% max





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • A comparison of free amino nitrogen and yeast assimilable nitrogen measurement methods for use in alcoholic fermentation of whey.: This research demonstrates the utility of ninhydrin in evaluating nitrogen sources for yeast fermentation, contributing significantly to dairy science and fermentation technology advancements (Schaan & Hughes, 2024).

  • Further research into alternative carrier solvents for the detection of latent fingermarks.: Explores ninhydrin′s application in forensic science, particularly in developing new solvent systems for enhancing the visibility of latent fingerprints, crucial for law enforcement and forensic investigations (Able et al., 2024).

  • Substrate-controlled [4+1] and [3+2] annulations of ninhydrin-derived Morita-Baylis-Hillman carbonates to access polysubstituted furans and cyclopentenes.: Highlights the versatility of ninhydrin in organic chemistry, providing new methodologies for synthesizing complex organic compounds, which can be pivotal for developing new pharmaceuticals and materials (Wang et al., 2024).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service