Skip to Content
Merck
All Photos(1)

Key Documents

MXPHUBKIT-M

Millipore

Milliflex® PLUS Pump VHP hub Kit

The Milliflex® PLUS Pump VHP hub kit is used for easy fluidic and electrical connection of the Milliflex PLUS Pumps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.79
Pricing and availability is not currently available.

Quality Level

manufacturer/tradename

Milliflex®

input

sample type pharmaceutical(s)

application(s)

bioburden testing
pharmaceutical
sterility testing
water monitoring

compatibility

for use with MILLIFLEX®
standard for VHP

shipped in

ambient

Components

Includes: 1 electrical hub and 3 cables

1 fluidic hub and 3 tubes

Power supply

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service