Skip to Content
Merck
All Photos(1)

Key Documents

MXAC00001

Millipore

Milliflex stainless steel filter support for manifold

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing, Water Monitoring

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24
Pricing and availability is not currently available.

material

stainless steel

Quality Level

manufacturer/tradename

Milliflex®

input

sample type bottled water
sample type beer
sample type cosmetics
sample type pharmaceutical(s)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

General description

Ready-to-use, sterile Milliflex filter units combine a funnel and a gridded membrane filter in one device.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service