Skip to Content
Merck

Skip To

MX0484

Methanol

≥99.5% (GC), suitable for GC/MS, OmniSolv®

Synonym(s):

Methyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
UNSPSC Code:
41116121
EC Index Number:
200-659-6
MDL number:
Beilstein/REAXYS Number:
1098229
Assay:
≥99.5% (GC)
Technique(s):
GC/MS: suitable
Bp:
64.7 °C (lit.)
Vapor pressure:
128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Methanol, pesticide residue grade OmniSolv®

vapor density

1.11 (vs air)

Quality Segment

vapor pressure

128 hPa ( 20 °C), 410 mmHg ( 50 °C), 97.68 mmHg ( 20 °C)

product line

OmniSolv®

assay

≥99.5% (GC)

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

GC/MS: suitable

impurities

≤0.05% Water (H2O), ≤0.3 μeq/g Titrable acid, ≤10 ppm Aldehydes and ketones

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Application

pesticide residue grade OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
is packaged in 1L amber glass bottle
is packaged in 4L amber glass bottle

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Aldehydes and ketones10ppm maxAssay (GC)99.5% minCap. ECD responsive substances as PCNB (peak)5ng/L maxCap. ECD responsive substances as PCNB (total)50ng/L maxCap. FID responsive substances as decane (peak)2µg/L maxCap. FID responsive substances as decane (total)20µg/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardTitrable acid0.3µeq/g maxWater (H2O)0.05% max
Evaporation Number: 1.9

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Methanol


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters