Skip to Content
Merck
All Photos(1)

Documents

MTRACECR1

Millipore

M-TRACE® Charging Cradle

for use with (M-TRACE® Touch Mobile Computer)

Synonym(s):

Charger

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
43210000
NACRES:
JA.12

packaging

pack of -1 unit

application(s)

pharmaceutical
sterility testing

compatibility

Steritest® Symbio pump
for use with (M-TRACE® Touch Mobile Computer)

General description

The M-Trace® Software & Mobile App is your 21 CFR Part 11 compliant Digital QC companion for sterility testing and other regulated processes, ensuring trusted and efficient data recording. Click here to get more details on the overall M-Trace® solution.

M-TRACE® Charging Cradle is a cleanroom-friendly accessory of the M-TRACE® Touch Mobile Computer (MTRACEMC1 ) and is compliant with the QC sterility testing environment requirements. The M-TRACE® Charging Cradle enables fast charging and storage of the M-TRACE® Touch Mobile Computer. Whenever the M-TRACE® Touch Mobile Computer is not in use, the device can be easily put to hold on the M-TRACE® Charging Cradle assuring safe storage and automatic recharge.

Application

M-TRACE® Charging Cradle enables fast charging and storage of the M-TRACE® Touch Mobile Computer during sterility testing and other quality control workflows

Legal Information

M-TRACE is a registered trademark of Merck KGaA, Darmstadt, Germany
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service