Skip to Content
Merck
All Photos(1)

Key Documents

MSSLBPC

Millipore

Multiscreen® 96 well Plate, polycarbonate membrane

pore size 0.4 μm, non-sterile

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€582.00
50 PKG
€2,460.00

€582.00


Estimated to ship on10 May 2025



Select a Size

Change View
10 EA
€582.00
50 PKG
€2,460.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

€582.00


Estimated to ship on10 May 2025


material

flat bottom wells
polycarbonate membrane
polystyrene

Quality Level

description

This clear 96 well filter plate has a Polycarbonate membrane used for Aqueous solubility assays. Comes in a pack of 10.

sterility

non-sterile

product line

MultiScreen®

feature

lid

manufacturer/tradename

MultiScreen®

parameter

50-250 μL sample volume (per well)

technique(s)

solubility assay: suitable

plate size

96 wells

well maximum volume

300 μL

working volume

50-250 μL

pore size

0.4 μm pore size

binding type

Tissue Culture (TC)-treated surface

shipped in

ambient

General description

Filter Code: PC
No matter what application you’re working with, Millipore has a plate developed to meet your specific needs. MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.

HTS Filter Plates (Plate catalogue numbers that start with MS)
- MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.

- Millipore’s new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.

Classic Filter Plates (Plate catalogue numbers that start with MA)
- The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.

Accessory – MultiScreenHTS Vacuum Manifold
- The MultiScreenHTS Vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.

Millicell Assay Plates
- In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.

Application

Research Category
Nucleic acid

Physical form

Color Code: Clear

Other Notes

Automation Compatible: Yes

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This provides information about the chemical compatibility of MultiScreen Filter Plates with different reagents.

Questions

  1. Do you have the product like this with 0.4 um pore size of polycarbonate membrane and be sterile?

    1 answer
    1. There are several product options available.
      Please see the links below to review possible sterile plate/insert products - including sterile, 0.4 µm polycarbonate membranes - that may be of interest:
      Millicell Inserts
      https://www.sigmaaldrich.com/substance/millicellstandingcellcultureinserts1234598765

      Transwell Insert:
      https://www.sigmaaldrich.com/US/en/substance/corninghtstranswell96wellpermeablesupports1234598765

      Other products in other formats are also available. To discuss this further, we kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice and click on "Product Technical Inquires" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service