MSHABGM48
MicropreSure® Filtration Unit
0.45 μm black gridded MCE membrane filter, bulk packaging, suitable for liquid cultivation
Synonym(s):
For bioburden and water analysis by filtration, MicropreSure® Monitor
About This Item
Recommended Products
material
black filter
cellulose support (pad)
gridded filter
mixed cellulose esters (MCE)
polycarbonate
Quality Level
description
packed
sterility
sterile; ethylene oxide treated
manufacturer/tradename
MicropreSURE®
filter diam.
47 mm
filtration area
9.1 cm2
matrix
MF-Millipore™
pore size
0.45 μm pore size
input
sample type dairy
sample type wine
sample type beer
sample type cider
sample type soft drink
sample type sport drink
sample type bottled water
sample type pharmaceutical(s)
sample type beverage(s)
sample type food(s)
application(s)
bioburden testing
food and beverages
industrial qc (on-line monitoring)
pharmaceutical
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Closed design for accurate sampling
- Fast and reliable in-process collection and microbiological analysis
- Tests larger volumes
- Batch or continuous sampling
- No assembly or sterilization needed
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
MicropreSure® on-line filtration system for in-process microbiology testing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service