Skip to Content
Merck
All Photos(1)

Key Documents

MR-006

Sigma-Aldrich

EmbryoMax® Modified DPBS with BSA and phenol red

liquid, with BSA and phenol red, EmbryoMax®

Synonym(s):

PBS, phosphate buffered saline

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€57.40

€57.40


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€57.40

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

€57.40


Estimated to ship on17 May 2025


Request a Bulk Order

Product Name

EmbryoMax® Modified DPBS with BSA and phenol red, The EmbryoMax Modified DPBS with BSA & phenol red is available in a 100 mL format and has been optimized and validated for Embryo Culture.

Quality Level

form

liquid

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | embryo: suitable
cell culture | stem cell: suitable

input

sample type: mouse embryo(s)

Storage and Stability

Store at 2 to 8°C for up to 2 months & 1 day

Note: Our EmbryoMax Liquid Mouse Embryo Media is produced on a bi-monthly basis. For regular users, we encourage standing orders to ensure that backorders do not occur.

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service