MCEP06H48
Millicell® Hanging Cell Culture Insert, PET 8 μm, 6-well, 48/pk
Discontinued with Replacement Product No. PTEP06H48
Synonym(s):
cell culture inserts, permeable culture insert, plate inserts, tissue culture insert, tissue culture plate
About This Item
Recommended Products
material
PET membrane
flat wells
polystyrene housing
sterility
sterile
feature
lid: no
packaging
pack of 48 ea (individually blister packaged)
parameter
50 °C max. temp.
filter diam.
24 mm
filtration area
4.5 cm2
size
6 wells
surface area
4.5 cm2
well maximum volume
5.9 mL
working volume
2 mL
pore size
8.0 μm
input
sample type cardiac stem cell(s)
sample type: human embryonic stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type pancreatic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type epithelial cells
sample type neural stem cell(s)
sample type mesenchymal stem cell(s)
sample type hematopoietic stem cell(s)
binding type
Tissue Culture (TC)-treated surface
General description
Millicell® Hanging Inserts: Preloaded in receivers or available standalone
- For co-culturing and permeability assays
- Unique design allows easier basolateral access than other hanging inserts with less risk of contamination
- Available in 5 pore sizes and 3 diameters, including a 1 μm pore size that is optically transparent for better visualization by microscopy
- Available preloaded in 24-well receiver plates or as individual inserts in 3 diameters
Applications: Cell Attachment, Cell Growth, Cell Differentiation, Immunocytochemistry
Application
Legal Information
replaced by
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This protocol covers 3 modes for the microscopic examination of cell samples.
This page covers the indirect co-culture of embryonic stem cells with embryonic fibroblasts.
This page covers the ECM coating protocols developed for four types of ECMs on Millicell®-CM inserts, Collagen Type 1, Fibronectin, Laminin, and Matrigel.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service