Skip to Content
Merck

Skip To

MAP2MAG-76K

MILLIPLEX® Mouse Acute Phase Panel 2

Configurable Mouse Acute Phase 6-Plex Panel 2

Synonym(s):

acute phase proteins, immune response proteins, immunoassay targets, inflammation-related analytes, inflammatory biomarkers, mouse cytokine markers, mouse inflammation markers, plasma proteins, preclinical inflammation indicators, serum biomarkers

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84
eCl@ss:
32161000
To order a Milliplex® kit, please search for your analyte of interest.


Product Name

MILLIPLEX® Mouse Acute Phase Panel 2,

Quality Segment

description

Configurable Mouse Acute Phase 6-Plex Panel 2

product line

Milliplex®

species reactivity

mouse

packaging

pkg of 1 ea

technique(s)

multiplexing: suitable

input

plasma
tissue/cellular extract(s)
cell culture supernatant(s)
serum

assay range

accuracy: 99.2-128.2%, sensitivity: 0.001-7.495 ng/mL, standard curve range: 0.001-0.5 ng/mL
(AGP), standard curve range: 0.005-4.0 ng/mL
(Adipsin), standard curve range: 0.014-10 ng/mL
(CRP), standard curve range: 0.549-400 ng/mL
(Haptoglobin), standard curve range: 13.717-10,000 ng/mL
(SAP), standard curve range: 6.858-5000 ng/mL
(A2M), inter-assay cv: <15%
intra-assay cv: <10%

detection method

fluorometric (Luminex® xMAP® technology)

shipped in

wet ice

storage temp.

2-8°C

General description

Metabolic hormones such as insulin, leptin, ghrelin, adiponectin, and resistin are central to regulating energy balance, appetite, and glucose metabolism. Insulin promotes cellular glucose uptake, leptin signals satiety, and ghrelin stimulates hunger, while adiponectin enhances insulin sensitivity and resistin contributes to inflammation and insulin resistance. Their coordinated activity reflects the body’s metabolic state and is crucial for understanding disorders like obesity and diabetes.

Application

MILLIPLEX® Qualified assays undergo rigorous assay development, verification, and Quality Control testing to achieve optimal performance. Simultaneously analyze up to 6 analytes in mouse serum, plasma, or cell culture supernatants.

Analytes included: α-1-Acid Glycoprotein (AGP), α-2-Macroglobulin, Adipsin (Factor D), C-Reactive Protein (CRP), Haptoglobin, Serum Amyloid P (SAP).

Assay Characteristics: Refer to assay protocol for details on cross-reactivity, sensitivity, precision, and accuracy.

Features and Benefits

Features:

  • Measures key metabolic hormones involved in energy regulation and glucose metabolism
  • Includes analytes such as insulin, leptin, ghrelin, adiponectin, and resistin
  • Enables simultaneous profiling for a comprehensive metabolic overview
  • Applicable to various biological samples like serum, plasma, and tissue extracts - Suitable for both research and clinical applications
Benefits:

  • Detects early signs of metabolic dysfunction such as insulin resistance and obesity
  • Supports evaluation of therapeutic interventions targeting metabolic pathways
  • Enhances understanding of hormonal interactions in metabolic disorders
  • Provides valuable data for tracking disease progression and recovery
  • Facilitates personalized approaches to managing metabolic health

Legal Information

Luminex is a registered trademark of Luminex Corporation
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

For research use only. Not for use in diagnostic procedures.




Label License/Sticker for Assay Product:

By opening the packaging containing this Assay Product (which contains fluorescently labeled microsphere beads authorized by Luminex Corporation) or using this Assay Product in any manner, you are consenting and agreeing to be bound by the End User Terms and Conditions and the End User License Agreement available at http://support.diasorin.com/end-user-terms-and-conditions/. If you do not agree to all of the terms and conditions, you must promptly return this Assay Product for a full refund prior to using it in any manner.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABMAG83KMKI1MAG94KMADKMAG71K
description

-

description

-

description

-

description

-

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

assay range

accuracy: 99.2-128.2%, standard curve range: 0.001-0.5 ng/mL
(AGP), standard curve range: 0.014-10 ng/mL
(CRP), standard curve range: 13.717-10,000 ng/mL
(SAP), inter-assay cv: <15%
intra-assay cv: <10%, standard curve range: 0.005-4.0 ng/mL
(Adipsin), standard curve range: 6.858-5000 ng/mL
(A2M), sensitivity: 0.001-7.495 ng/mL, standard curve range: 0.549-400 ng/mL
(Haptoglobin)

assay range

accuracy: 80%
(Spike recovery for Aβ1-40), sensitivity: 3.6 pg/mL
(A?1-40; MinDC+2SD), sensitivity: 4.8 pg/mL
(A?1-42; MinDC+2SD), standard curve range: 3-2500 pg/mL
(Aβ1-40), standard curve range: 7-5000 pg/mL
(Aβ1-42), inter-assay cv: <10%
intra-assay cv: <5%

assay range

accuracy: 73-98%, sensitivity: 0.001-0.041 ng/mL
(MinDC+2SD), standard curve range: 0.001-1 ng/mL
(VEGF), standard curve range: 0.005-5 ng/mL
(IP-10), standard curve range: 0.01-15 ng/mL
(KIM-1), standard curve range: 0.02-25 ng/mL
(TIMP-1), standard curve range: 0.05-50 ng/mL
(β-2-Microglobulin), standard curve range: 0.05-50 ng/mL
(Renin)

assay range

accuracy: 77-92%, standard curve range: 12.2-50,000 pg/mL

manufacturer/tradename

Milliplex®

manufacturer/tradename

Milliplex®

manufacturer/tradename

Milliplex®

manufacturer/tradename

Milliplex®

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

target_organs

Respiratory Tract

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters