Skip to Content
Merck

Skip To

MACL09600

MultiScreen Column Loader

100µL, Black, Non-Sterile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41104923
NACRES:
NB.22
eCl@ss:
32039006
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

aluminum (with non-stick, non-abrasion coating)

Quality Segment

sterility

non-sterile

product line

MultiScreen®

usage

Manual

packaging

pkg of 1 ea

technique(s)

activity assay: suitable (protein kinase/phosphatase), bead-based assay: suitable, ligand binding assay: suitable, receptor binding assay: suitable

plate size

96 wells

working volume

100 μL

application(s)

sample preparation

shipped in

ambient

storage temp.

no temp limit

General description

No matter what application you’re working with, Millipore has a plate developed to meet your specific needs. MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.

HTS Filter Plates (Plate catalogue numbers that start with MS)
- MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.

- Millipore’s new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.

Classic Filter Plates (Plate catalogue numbers that start with MA)
- The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.

Accessory – MultiScreenHTS Vacuum Manifold
- The MultiScreenHTS Vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.

Millicell Assay Plates
- In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.

Application

100µL, Black, Non-Sterile
Research Category
Assays

Physical form

Color Code: Black

Other Notes

Automation Compatible: No

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSVMHTSHVMAMP09608MACF09604
description

100µL, Black, Non-Sterile

description

Vacuum Filtration

description

Radionucleotide counting by transfer of individual filters to tubes

description

This Centrifuge Alignment Frame is used to align MultiScreen plate to 96-well receiver plate for centrifugation of aqueous media.

packaging

pkg of 1 ea

packaging

pkg of 1 ea

packaging

pkg of 1 ea

packaging

pkg of 4 ea

material

aluminum (with non-stick, non-abrasion coating)

material

Nylon

material

-

material

blue acrylonitrile-butadiene-styrene (ABS) plate

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

storage temp.

no temp limit

usage

Manual

usage

Automated & Manual

usage

Manual

usage

Manual

product line

MultiScreen®

product line

MultiScreen®

product line

-

product line

MultiScreen®


Still not finding the right product?

Explore all of our products under MultiScreen Column Loader




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters