Skip to Content
Merck
All Photos(1)

Key Documents

MAC131M

Sigma-Aldrich

Mouse anti-Camelid IgG2/3 (heavy chain) Antibody, clone P17JE11

clone P17JE11, from mouse

Synonym(s):

HCAbs, Camelid antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

P17JE11, monoclonal

species reactivity

Camelidae, camel, llama

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable

isotype

IgG1κ

target post-translational modification

unmodified

Related Categories

General description

Camelidae family that includes camels and llamas are the only mammals reported to have heavy-chain (IgG-like) antibodies. Camelids produce conventional antibodies, but they also produce antibodies that do not associate with light chains. They produce functional antibodies devoid of light chains of which the single N-terminal domain is fully capable of antigen binding. Their VHH domain is located on a single stretch of amino acids and retains its antigen-recognition capability. Their heavy chains are devoid of the CH1 domain and have modified VH domain. Camelid heavy-chain antibodies (HCAbs) comprise about 50% of serum IgG in Llama and about 75% in Camelus species. HCAbs exhibit a molecular mass of about 100 kDa, which is smaller than conventional IgG1 and allows it to bind epitopes that may not be accessible to conventional antibodies. Also, the smaller size and single domain make it easier to transform them into bacterial cells for larger production needs. HCAbs are reported to display similar specificities, but are more robust in some cases. (Ref.: Daley LP et al. (2010). Clin. Vaccine Immunol. 17(2); 239-246; Harmsen, MM., and De Haard, HJ (2007) Appl Microbiol Biotechnol 77(1):13-22).

Specificity

Clone P17JE11 detects Camelid IgG2/igG3 heavy chain. Displays trivial to none reactivity with other species.

Immunogen

Camelid heavy-chain immunoglobulin G (clgG2/3).

Application

Mouse anti-Camelid IgG2/3 (heavy chain), clone P17JE11 detects Camelid IgG2/igG3 heavy chain. It has been tested for use in ELISA.
Research Sub Category
Secondary & Control Antibodies

Quality

Evaluated by ELISA in IgG as tested in various species.

ELISA Analysis: 2 µg/mL of this antibody detected Camelid IgG2/3 (heavy chain).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service