Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
P17IG12, monoclonal
Species reactivity:
mouse, Camelidae
Application:
ELISA
Citations:
-
antibody form
purified antibody
Quality Segment
clone
P17IG12, monoclonal
purified by
using protein G
species reactivity
mouse, Camelidae
concentration
(Please refer to lot specific datasheet.)
technique(s)
ELISA: suitable
isotype
IgG2aκ
General description
Polyclonal Camelid IgG2a (long-hinge hcAb corresponding to Camelid (total).
Physical form
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by ELISA in HeLa cell lysate.
ELISA Analysis: 1.0 µg/mL of this antibody detected Llama IgG1, llama IgG2, and llama IgG3 in ELISA.
ELISA Analysis: 1.0 µg/mL of this antibody detected Llama IgG1, llama IgG2, and llama IgG3 in ELISA.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.