Skip to Content
Merck
All Photos(1)

Key Documents

MABX38

Sigma-Aldrich

Anti-Norovirus genogroup I

Synonym(s):

Anti-Norovirus Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

MABX38-1KC
€752.00
MABX38-10KC
€6,010.00

€752.00


Estimated to ship on19 April 2025


Request a Bulk Order

Select a Size

Change View
MABX38-1KC
€752.00
MABX38-10KC
€6,010.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€752.00


Estimated to ship on19 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

monoclonal

form

solution

species reactivity

all

packaging

pkg of 1 mg (MABX38-1KC)
pkg of 10 mg (MABX38-10KC)

technique(s)

ELISA: suitable
immunoblotting: suitable
western blot: suitable

isotype

IgG2a

relevant disease(s)

infectious diseases

shipped in

wet ice

storage temp.

2-8°C

Specificity

Group I capsid protein. No reactivity with genogroup II

Immunogen

Recombinant Norovirus VLP

Application

This mouse monoclonal , Cat. No. MABX38, is tested for use in ELISA, Lateral Flow Assay, and Western Blotting, for the detection of Group I capsid protein.

Target description

Noroviruses are a genetically diverse positive stranded RNA virus of the Caliciviridae family. Genotypes I, II and IV can cause the sudden onset of sever vomiting and diarrhea.  The virus is highly contagious and commonly spread through contaminated food, water and through close contact with infected persons.

Linkage

Recommended Matched Pair MABX38 capture and MABX39 detection

Physical form

Purified mouse monoclonal in buffer containing 10 mM Phosphate Buffered Saline, pH 7.2 with 0.1% sodium azide.

Reconstitution

Please refer to Certificate of Analysis for concentratio

Storage and Stability

Store at 2-8 °C. For long-term storage, freeze at -20 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service