Skip to Content
Merck
All Photos(4)

Documents

MABT903

Sigma-Aldrich

Anti-Connexin 43 Antibody, N-terminal Antibody, clone P1E11.B19

clone P1E11, from mouse

Synonym(s):

Gap junction alpha-1 protein, CX43, Gap junction 43 kDa heart protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P1E11, monoclonal

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rabbit (based on 100% sequence homology), horse (based on 100% sequence homology), canine (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology), chicken (based on 100% sequence homology), hamster (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

bovine ... Gja1(281193)
chicken ... Gja1(395278)
dog ... Gja1(403418)
horse ... Gja1(100067229)
human ... GJA1(2697)
mouse ... Gja1(14609)
rabbit ... Gja1(100008935)
rat ... Gja1(24392)
rhesus monkey ... Gja1(714344)

General description

Gap junction alpha-1 protein (UniProt: P08050; also known as Connexin-43, CX43, Gap junction 43 kDa heart protein) is encoded by the Gja1 (also known as Cxn-43) gene (Gene ID: 24392) in rat. The gap junction channels are dodecamers of transmembrane proteins of the connexin family. Connexins usually have a short half-life ( 1.5 - 5 h) and this short half-life is a key element of coupling regulation as it allows very dynamic and acute changes in gap junction regulation and gap junctional communication. In cardiomyocytes three types of connexins have been identified: Connexin 40, 43, and 45. Each of these connexins form channels with unique and specific electrophysiological properties. Connexin-43 is a multi-pass membrane protein and is the most ubiquitous connexin with widespread tissue expression pattern. Connexin-43 can be phosphorylated at multiple sites and phosphorylation is shown to regulate the kinetics of its trafficking, assembly, gating and turnover in a cell-cycle-stage specific manner. Phosphorylation at Ser325, Ser328 and Ser330 by casein kinase I is reported to modulate gap junction assembly while phosphorylation of Ser368 by protein kinase C gamma isoform leads to disassembly of gap junction plaques and inhibition of gap junction activity. During myocardial ischemia connexin-43 is shown to become dephosphorylated that allows gap junctions to be opened and convey the propagation of ischemia injury. In bladder smooth muscle cells, connexin-43 displays a circadian cycle with low levels during the sleep phase.

Specificity

Clone P1E11.B19 detects connexin-43 in most mammalian species. It targets an epitope with in the first 20 amino acids from the N-terminal end.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the first 20 amino acids from the N-terminal region of rat connection-43.

Application

Research Category
Cell Structure
This mouse monoclonal Anti-Connexin 43, N-terminal, clone P1E11.B19, Cat. No. MABT903 is tested for use in Immunocytochemistry and Western Blotting for the detection of Connexin-43.
Western Blotting Analysis: A representative lot detected Connexin 43 in Western Blotting applications (Sosinsky, G.E., et. al. (2007). Biochem. 408(3):375-85).

Western Blotting Analysis (WB): A representative lot detected Connexin 43 in NRK cells (Courtesy of Joell Solan in Paul Lampe s Lab at Fred Hutchinson Cancer Research Center, Seattle, WA).

Western Blotting Analysis: A representative lot detected Connexin 43 in Western Blotting applications (Kalcheva, N., et. al. (2007). Proc Natl Acad Sci USA. 104(51):20512-6).

Western Blotting Analysis: 1-4 µg/mL from a representative lot detected Connexin 43 in 10 µg of rat and mouse heart tissue lysates.

Immunocytochemistry Analysis: A representative lot detected Connexin 43 in Immunocytochemistry applications (Gellhaus, A., et. al. (2004). J Biol Chem. 279(35):36931-42).

Western Blotting Analysis: A representative lot detected Connexin 43 in Western Blotting applications (Goldberg, G.S., et. al. (2002). J Biol Chem. 277(39):36725-30).

Western Blotting Analysis: A representative lot detected Connexin 43 in Western Blotting applications (Solan, J.L., et. al. (2003). J Cell Sci. 116(Pt 11):2203-11).

Quality

Evaluated by Western Blotting in human heart tissue lysates.

Western Blotting Analysis: 1 µg/mL of this antibody detected Connexin 43 in 10 µg of human heart tissue lysates.

Target description

~43 kDa observed; 43.03 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinbo Li
International journal of physiology, pathophysiology and pharmacology, 11(5), 212-219 (2019-11-30)
Intercellular communication via gap junctions and tunneling nanotubes (TNT) play pivotal roles for cell differentiation and proliferation and homeostasis. Intercellular communication has been found in human trabecular meshwork cells, which are key elements for allowing the direct passing aqueous humor
Jonathan Boucher et al.
Cancers, 12(10) (2020-10-22)
Among the different interacting molecules implicated in bone metastases, connexin43 (Cx43) may increase sensitivity of prostate cancer (PCa) cells to bone microenvironment, as suggested by our in silico and human tissue samples analyses that revealed increased level of Cx43 expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service