Skip to Content
Merck
All Photos(2)

Key Documents

MABT38

Sigma-Aldrich

Anti-Laminin subunit alpha-4 Antibody, clone 3H2

clone 3H2, from mouse

Synonym(s):

laminin, alpha 4, laminin subunit alpha-4, Laminin-8 subunit alpha, Laminin-9 subunit alpha, laminin alpha 4 chain, Laminin-14 subunit alpha

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€379.00

€379.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€379.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3H2, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LAMA4(3910)

General description

Laminins are basement membrane, extracellular matrix glycoproteins that are composed of alpha, beta and gamma heterotrimeric chains. Different combinations of five alpha chains, three beta chains, and two gamma chains form at least 12 laminin isoforms. Laminin subunit alpha-4 contains the C-terminal G domain, and although implicated in developmental regulation due to expression profiles, the biological function of laminin subunit alpha-4 is yet to be fully understood. Laminis as a whole play an important role in cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. High expression has been observed in adult heart, lung, ovary, small and large intestines, placenta, and liver, with lower levels observed in skeletal muscle, kidney, pancreas, testis, prostate, and rain.

Immunogen

Recombinant protein corresponding to human Laminin subunit alpha-4.

Application

Anti-Laminin subunit alpha-4 Antibody, clone 3H2 detects level of Laminin subunit alpha-4 & has been published & validated for use in WB, IH, IP, ELISA.
Immunohistochemistry Analysis: A 1:500 dilution from a previous lot detected Laminin subunit alpha-4 in basal cell carcinoma tissue.

ELISA Analysis: A previous lot was used by an independent laboratory in ELISA. (Wondimu, Z., et al. (2004). Blood. 104(6):1859-1866.)

Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP. (Wondimu, Z., et al. (2004). Blood. 104(6):1859-1866.)
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Evaluated by Western Blot in human serum tissue lysate under non-reduced conditions.

Western Blot Analysis: A 1:50,000 dilution of this antibody detected Laminin subunit alpha-4 in 10 µg of human serum tissue lysate (non-reduced conditions).

Target description

~202 kDa observed under non-reduced conditions.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human serum tissue lysate (non-reduced)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service