Skip to Content
Merck
All Photos(1)

Key Documents

MABT213

Sigma-Aldrich

Anti-SHP-1 Antibody, clone HG213, rabbit monoclonal

clone HG213, from rabbit, unconjugated

Synonym(s):

Tyrosine-protein phosphatase non-receptor type 6, Hematopoietic cell protein-tyrosine phosphatase, Protein-tyrosine phosphatase 1C, PTP-1C, Protein-tyrosine phosphatase SHP-1, SH-PTP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€446.00

€446.00


Estimated to ship on07 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on07 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

unpurified

antibody product type

primary antibodies

clone

HG213, monoclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPN6(5777)

General description

Src Homology region 2 domain Phosphatase 1 (SHP-1) is also known as Protein Tyrosine Phosphatase 1C (PTP1C), Protein Tyrosine Phosphatase non-receptor type 6 (PTPN6), and Hematopoietic Cell Phosphatase (HCPH). SHP-1 is a SH2 domain-containing cytosolic tyrosine phosphatase characterized by a catalytic phosphatase domain of approximately 240 amino acids that selectively dephosphorylates tyrosine residues in proteins; the SH2 domains are critical for SHP-1 to bind to ITIM domains in inhibitory immunoreceptors. SHP-1 is highly expressed in hematopoietic tissues and has a negative impact on lymphocyte signaling. SHP-1 is implicated in the control of tyrosine kinase signaling pathways in cellular proliferation, with a potential role in cancer.

Immunogen

Full-length glutathione transferase SHP-1 fusion protein, purified from bacteria.

Application

Anti-SHP-1 Antibody, clone HG213 detects level of SHP-1 & has been published & validated for use in WB.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected SHP-1 in 10 µg of Jurkat cell lysate.

Target description

~67 kDa observed.
3 isoforms exist with molecular weights between 63 kDa and 68 kDa

Physical form

Rabbit monoclonal supernatant without sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Jurkat cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service