Skip to Content
Merck
All Photos(2)

Key Documents

MABT163

Sigma-Aldrich

Anti-γ-Tubulin Antibody

mouse monoclonal, 6H3.1

Synonym(s):

Tubulin gamma-1 chain, Gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

Anti-Gamma-1-Tubulin, clone 6H3.1 Antibody, clone 6H3.1, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6H3.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TUBG1(7283)

General description

Gamma-1-Tubulin, also known as Gamma-tubulin complex component 1, GCP-1 or Tubulin gamma-1 chain, and encoded by the gene TUBG1/TUBG, is a form of tubulin protein and a component of microtubules. Tubulin is the major constituent of microtubules. The gamma chain is found at microtubule organizing centers (MTOC) such as the spindle poles or the centrosome and plays a major role in the organization of the microtubule cytoskeleton. Gamma-1-Tubulin is part of the pericentriolar matrix component that regulates alpha/beta chain minus-end nucleation, and it is also involved in centrosome duplication and spindle formation. Interestingly, gamma tubulin is often up regulated but improperly compartmentalized in many cancer cells including non-small cell lung cancer. Mutations in Gamma-1-Tubulin gene are associated with developmental cortical defects including microcephaly and malformations of cortical development (MCD). Gamma-1-Tubulin is localized to the cytoplasm and cytoskeleton components and expressed in a variety of cell types and tissues.

Immunogen

GST-tagged recombinant protein corresponding to human Gamma-1-Tubulin.

Application

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected Gamma-1-Tubulin in NIH/3T3 cells.

Alexa Fluor is a registered trademark of Life Technologies.
This Anti-Gamma-1-Tubulin antibody, clone 6H3.1 is validated for use in western blotting & ICC for the detection of Gamma-1-Tubulin.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected Gamma-1-Tubulin in 10 µg of HeLa cell lysate.

Target description

~56 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jens S Andersen et al.
Nature communications, 15(1), 2687-2687 (2024-03-28)
Centrosomes and cilia are microtubule-based superstructures vital for cell division, signaling, and motility. The once thought hollow lumen of their microtubule core structures was recently found to hold a rich meshwork of microtubule inner proteins (MIPs). To address the outstanding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service