Skip to Content
Merck
All Photos(1)

Key Documents

MABT1495

Sigma-Aldrich

Anti-PD2 (PAF1) Antibody, clone F169-3B2

clone F169-3B2, from mouse

Synonym(s):

Pancreatic differentiation protein 2, RNA polymerase II-associated factor 1 homolog, hPAF1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€116.00
100 μG
€315.00

€116.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
€116.00
100 μG
€315.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€116.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F169-3B2, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PAF1(54623)

General description

Pancreatic differentiation protein 2 (UniProt: Q8N7H5; also known as RNA polymerase II-associated factor 1 homolog, PD2, hPAF1) is encoded by the PAF1 (also known as PD2) gene (Gene ID: 54623) in human. PD2 is a nuclear protein that serves as a subunit of the RNA polymerase II associated factor (PAF) complex. It has multiple functions during transcription by RNA polymerase II. It has been implicated in regulation of development and maintenance of embryonic stem cell pluripotency. It associates with RNA polymerase II through interaction with POLR2A C-terminal domain non-phosphorylated and serine 2 and serine 5 phosphorylated forms. It is involved in transcriptional elongation, acting both independently and synergistically with TCEA1 and in cooperation with the DSIF complex and HTATSF1. Three isoforms of PD2 have been described that are produced by alternative splicing. PD2 has oncogenic property and is shown to be overexpressed in pancreatic cancer cells. In normal murine pancreas, its expression is restricted to acinar cells, however, its expression is increased in the ductal cells of mouse model of pancreatic cancer with advancing age. (Ref.: Dey, P., et al (2014). Oncotarget, 5(12); 4480-4491).

Specificity

Clone F169-3B2 detects Pancreatic differentiation protein 2 (PD2) in human. It targets an epitope with in 22 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal half of human Pancreatic differentiation protein 2 (PD2).

Application

Anti-PD2 (PAF1), clone F169-3B2, Cat. No. MABT1495, is mouse monoclonal antibody that detects Pancreatic differentiation protein 2 and has been tested for use in ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected PD2 (PAF1) in Immunocytochemistry applications (Dey, P., et. al. (2014). Oncotarget. 5(12):4480-91; Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).

Immunoprecipitation Analysis: A representative lot immunoprecipitated PD2 (PAF1) in Immunoprecipitation applications (Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).

ELISA Analysis: A representative lot detected PD2 (PAF1) in ELISA applications (Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).

Immunohistochemistry Analysis: A representative lot detected PD2 (PAF1) in Immunohistochemistry applications (Dey, P., et. al. (2014). Oncotarget. 5(12):4480-91).

Western Blotting Analysis: A representative lot detected PD2 (PAF1) in Western Blotting applications (Moniaux, N., et. al. (2006). Oncogene. 25(23):3247-57).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Immunocytochemistry in Panc1 cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected PD2 (PAF1) in Panc1 cells.

Target description

59.98 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service