Skip to Content
Merck
All Photos(1)

Key Documents

MABT1460

Sigma-Aldrich

Anti-LAMC2 Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€130.00
100 μL
€326.00

€130.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€130.00
100 μL
€326.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€130.00


Estimated to ship on27 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

P2H, monoclonal

mol wt

calculated mol wt 130.98 kDa

species reactivity

mouse, human

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... LAMC2(3918)

General description

Laminin subunit g2 (UniProt: Q13753; also known as Cell-scattering factor 140 kDa subunit, CSF 140 kDa subunit, Epiligrin subunit gamma, Kalinin subunit gamma, Kalinin/nicein/epiligrin 100 kDa subunit, Ladsin 140 kDa subunit, Laminin B2t chain, Laminin-5 subunit gamma, Large adhesive scatter factor 140 kDa subunit, Nicein subunit gamma) is encoded by the LAMC2 (also known as LAMB2T, LAMNB2) gene (Gene ID: 3918) in human. Laminin is a major basement membrane glycoprotein consisting of α, β, γ chains that are bound to each other by disulfide bonds. Over 15 different isoforms have been reported in the laminin family that are expressed in a tissue-specific manner and play different role in each tissue. γ2, a subunit of Laminin-5 is reported to be frequently over-expressed as a monomer or as the β2−γ3 heterodimer in invasive cancers and is regarded as an invasion marker. It has 8 EGF-like domains. The cell adhesion and motility activities of Laminin-5 are reported to be regulated by the proteolytic cleavage of the short arm of Laminin γ2 chain by BMP-1 or MT1-MMP. This cleavage releases a 45 kDa N-terminal proteolytic fragment (γ2pf). The short arm and γ2pf can be further cleaved by serine proteases and by matrix metalloproteinases resulting in the release of small N-terminal fragments. γ2pf-like fragments have been reported to be present in conditioned media of cultured cancer cells and in sera from human cancer subjects. Clone P2H targets the second N-terminal EGF-like repeat (NE2) of domain V of laminin 2. (Ref.: Sato, H., et al. (2014). Cancer Sci. 105(2); 168-175; Miyazaki, K., et al. (2016). Cancer Sci. 107(12); 1909-1918).

Specificity

Clone P2H is a mouse monoclonal antibody that detects Laminin-5, g2 chain. It targets an epitope within the N-terminal, NE2 in domain V.

Immunogen

His-tagged 45-kDa N-terminal fragment consisting of g2pf, domain V.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Laminin-5 subunit g2 (LAMC2) in A431 cells.

Tested Applications


Western Blotting Analysis: 1 μg/mL from a representative lot detected LAMC2 in lung squamous cell carcinoma cell lysates (Courtesy of Kaoru Miyazaki, Ph.D., KIhara Institute for Biological Research, Yokohama City University, Japan).

Western Blotting Analysis: A representative lot detected LAMC2 in Western Blotting applications (Miyazaki, K., et. al. (2016). Cancer Sci. 107(12):1909-1918).

Immunohistochemistry (Paraffin) Analysis: 10 μg/mL from a representative lot detected LAMC2 in normal human skin, skin cancer, normal breast, and breast adenocarcinoma tissues (Courtesy of Kaoru Miyazaki, Ph.D., KIhara Institute for Biological Research, Yokohama City University, Japan).

ELISA Analysis: 5 μg/mL from a representative lot detected LAMC2 in Lm-gamma2pf and Lm-gamma2 in Lm332 condition medium from Lm332-expressing HEK293 cells (Courtesy of Kaoru Miyazaki, Ph.D., KIhara Institute for Biological Research, Yokohama City University, Japan).

Immunocytochemistry Analysis: A representative lot detected LAMC2 in Immunocytochemistry applications (Miyazaki, K., et. al. (2016). Cancer Sci. 107(12):1909-1918).

Immunohistochemistry Applications: A representative lot detected LAMC2 in Immunohistochemistry applications of lung squamous cell carcinomas and lung adenocarcinomas (Courtesy of Kaoru Miyazaki, Ph.D., KIhara Institute for Biological Research, Yokohama City University, Japan).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-LAMC2, clone P2H, Cat. No. MABT1460, is a mouse monoclonal antibody that detects Laminin subunit g2 and is tested for use in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service