Skip to Content
Merck
All Photos(1)

Key Documents

MABT1444

Sigma-Aldrich

Anti-Tetranectin/CLEC3B Antibody, clone mAb10

Synonym(s):

Anti-CLEC3B Antibody, CLEC3B Detection Antibody, Clone mAb10 Anti-Tetranectin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€146.00
100 μG
€369.00

€146.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€146.00
100 μG
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

€146.00


Estimated to ship on15 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

mAb10, monoclonal

mol wt

calculated mol wt 22.54 kDa
observed mol wt ~25 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
dot blot: suitable
western blot: suitable

isotype

IgG2bκ

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CLEC3B(7123)

Related Categories

General description

Tetranectin (UniProt: P05452; also known as TN, C-type lectin domain family 3 member B, Plasminogen kringle 4-binding protein) is encoded by the CLEC3B (also known as TNA) gene (Gene ID: 7123) in human. Tetranectin is a tetrameric plasminogen-binding protein that is synthesized with a signal peptide (aa 1-21), which is subsequently removed in the mature form that is composed of three identical, noncovalently linked subunits each of 181 amino acids. The polypeptide consists of a short a-helical segment (aa 47-72) at the amino terminus that is involved in trimerization, followed by a C-type lectin domain (aa 77-198). It is present in both serum and the extracellular matrix and is expressed most abundantly in the lung. The blood concentrations of tetranectin in healthy adults are about 10-12 mg/mL. However, in septic subjects its levels are significantly reduced and supplementation with exogenous tetranectin confers dose-dependent protection against lethal endotoxemia and sepsis. Its levels are shown to be significantly reduced in patients with cardiac failure. Hence, it is also used as a biomarker for risk of heart failure. (Ref.: Chen, W., et al. (2020). Sci. Transl. Med. 12(539); eaaz3833; McDonald, K., et al. (2020). Sci. Rep. 10; 7507).

Specificity

Clone mAb10 is a mouse monoclonal antibody that detects Tetranectin. It targets an epitope within 24 amino acids from the C-terminal half.

Immunogen

Purified human Tetranectin.

Application

Quality Conrol Testing

Evaluated by Western Blotting in Human lung tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Tetranectin/CLEC3B in Human lung tissue lysate.

Tested Applications

Western Blotting Analysis: A representative lot detected Tetranectin/CLEC3B in Western Blotting applications (Chen, W., et al. (2020). Sci Transl Med. 12(539):eaaz3833).

Dot Blot: A representative lot detected Tetranectin/CLEC3B in Dot Blot applications (Chen, W., et al. (2020). Sci Transl Med. 12(539):eaaz3833).

ELISA Analysis: A representative lot detected Tetranectin/CLEC3B in ELISA applications (Chen, W., et al. (2020). Sci Transl Med. 12(539):eaaz3833).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Tetranectin/CLEC3B, clone mAb10, Cat. No. MABT1444, is a mouse monoclonal antibody that detects Tetranectin and is tested for use in Dot Blot, ELISA, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service